Stirling Ultracold ULT Freezers – Compressor free Technology for your Samples and Your World
Friday, 19th November, 2020
12:30 to 14:00 IST (Delhi, Kolkata, Chennai, Mumbai)
The purpose of our presentation is to provide insight on the compressor free technology of the Stirling engine & how its robust performance parameters are ideal for the environment & the storage of sample at any temperature from -20C to -86C. The performance characteristics of the Stirling engine allow for operation in higher ambient temperature conditions as well as areas with poor power quality.
The strength of the Stirling engine to remove constant amounts of heat, without performance degradation, also make the application ideal for areas with high freezer access or processes that require large amounts of ambient temperature products. We will review the environmentally sustainable attribute of the freezer, from low energy use to environmentally friendly refrigerants & our zero waste manufacturing factory. From a sample storage perspective we will present industry leading temperature uniformity & stability data to show how your samples & products are in the best possible environment.