Turnkey Solutions
VWR, a brand which serves global science industry with its complete range of products offers Turnkey Solution to help in buiding the facilities and its utilities.
We offer solutions under following sub-caegories.
- Conceptualization and Designing
- Civil Construction
- MEP Services
- Buiding Cleanroom
- Building Laboratories
- Furniture Supply & Installation
- Validation / Certification
- Supply of Instruments / Consumables / Chemicals
- Maintenance (AMCs)
Conceptualization and Designing
With the consultation of customer we build concept and design to suits your requirements and also adhear to all internation standard norms. Designing the facility / labs is very critical interms of utilisation of space which constitute major portion of the cost and easy movement of men, material and test samples. All internatinal standards calls for design standards to get accredited.
With experienced & specialised team of different industries we serve, will provide best in industry design for your critical activities which utilises available space and meet international standards with safety as a primary criteria.
Civil Construction
We take construction as a part of turnkey solution and works with renowned buiders who are well experienced in construction industry. Civil construction is big part of the turnkey it plays very important role in facility life, space utilisation etc., Working with us for the construction will give customer a real advantage of turnkey solution.
MEP Services
For effective & optimise functioning of any facility / lab depends on the Mechanical, Electrical, Plumbing & fire safety design and implementation. MEP is lifelne of any building and it is important they are developed with utmost accuracy and perfection. Our MEP design validation services provide sustainable solutions for projects across industries we work.
Our engineers with diverse skills and extensive domain expertise create MEPF BIM models and validate these models for constructability, performance, and maintenance of the building. Our familiarity with various codes such as IPC, ASHRAE, NFPA and the ability to follow Client/industry specific Drafting standards ensures flexible solutions for the customers and assures efficient deliveries.
Building Cleanroom
Require in manufacturing or scientific research, a cleanroom is a controlled environment that has a low level of pollutants such as dust, airborne microbes, aerosol particles, and chemical vapors. Cleanroom has a controlled level of contamination that is specified by the number of particles per cubic meter at a specified particle size.
A cleanroom is an investment and a real asset in a company’s strategy and needs to be well thought through. Many decisions must be taken when building a cleanroom, and these will affect the cleanroom performance, ergonomics and cost. We offer turnkey cleanroom engineering, design services and supply & buiding for most cleanroom projects.
We offer variety of cleanroom wall finishes including steel, aluminum, FRP, vinyl-covered gypsum, epoxy paint, emulsion paint, and hardboard. What type of finish chosen depends on the equipment to be used on them, cleanroom classification, usage, and product manufacturing. And finishes are carefully evaluated for specific compliance to meet building regulatory requirements and testing criteria as such ASTM E84 Flame Spread and Smoke Development Values and specific health and safety requirements and local seismic codes.
"Cleanrooms are classified according to the number and size of particles permitted per volume of air. Large numbers like ""class 100"" or ""class 1000"" refer to FED_STD-209E, and denote the number of particles of size 0.5 mm or larger permitted per cubic foot of air. The standard also allows interpolation, so it is possible to describe e.g. ""class 2000.""
Small numbers refer to ISO 14644-1 standards, which specify the decimal logarithm of the number of particles 0.1 µm or larger permitted per cubic metre of air. So, for example, an ISO class 5 cleanroom has at most 105 = 100,000 particles per m³.
Both FS 209E and ISO 14644-1 assume log-log relationships between particle size and particle concentration. For that reason, there is no such thing as zero particle concentration. Ordinary room air is approximately class 1,000,000 or ISO 9.
Building Laboratories
Building labs includes Wall partitions, Flooring, False Ceilining and Furnitures. whether it is a cleanroom or general lab it requires all the mentioned components to function.
We offer a complete range of cleanroom partition, ceiling systems and Flooring. These systems meet the technical requirements of high specification controlled environments. The ceiling panels are non-particle shedding, load bearing, walkable panels and feature pre-formed cut outs for the housing of HEPA filters and other dedicated services. Different types of walls like Freestanding, Glass Partitions and Cleanroom Wall Partitions with different material of construcctions can be supplied to meet the requirements.
Whether you’re designing a new lab, or renovating an old one, the toughest part is nailing down all of the tiny details. Flooring is one of the important thing to decide. Laboratory floors need to be slip-resistant, non-staining, non-permeable / non-absorbent / moisture-proof, durable, easy to clean and easy to maintain. We provide all types of flooring solution starting from Vetrified, ceramic, Viny etc.,
Furniture Supply & Installation
Laboratory with proper furniture makes a difference in terms of the work flow, safety, and operator efficiency and comfort.Lab benches, chairs, stools, tables, work surfaces, and storage cabinets are used in all types of facilities and laboratories.
We offers a wide variety of laboratory furniture to suit the needs of every lab. Fume hoods, laboratory tables, sink units, storage cupboards and other furniture can be purchased as standard units or individually modified according to the needs of the customer. We offer furniture is made of wood or of metal sheets, depending on the customer's preference for general Labs and SS furniture for cleanroom & controlled environments. Worktops and sinks can be chosen from a wide range of materials depending on the application & requirements. We also offers furniture for offices.
Validation / Certification
Validation is the important part of any facility / laboratory construction which enables customers to adhears to standards. We work with reputed third party validation / certification providers for validating the facilities as per the customer requirements. All international standards will be followed while validating / certifying the laboratories so that customers will get required accreditation for their work.
Supply of Instruments / Consumables / Chemicals
On completion of facility / laboratory supply of equipment / instrument from our core & market sourced range will be add on advanture for the customer. We take care of supply, installation & documentations of all the instruments we supply. Additionally we can also supply required chemicals, kits and consumables from our offers which makes customer to work with ONLY ONE supplier for all their regular requirements to run their facility / laboratories.
Maintenance (AMCs)
With the team of engineers who are qualified to repair / maintain the instruments we take annual maintenance (AMCs) of all the instrunets we supply also other instruments from other supplies after the completion of warranty. Thus makes the customer sure of running their facility/laboratories without any challenges.