Oxford Nanopore Technologies
The affordable, pocket-sized MinION and MinION Mk1C devices, which can deliver tens of gigabases of data in a single run, make sequencing accessible to any researcher across a wide range of applications.
Benefits of nanopore sequencing technology include:
- Any length read — from short to ultra-long (>4 Mb)
- Streamlined workflows — library prep in as little as 10 minutes
- Sequence native DNA/RNA — eliminate amplification-bias and directly identify epigenetic modifications (e.g. methylation)
- Real-time data streaming — immediate access to actionable results
- Portable, affordable devices — sequence anywhere
Sequencing devices
Get complete control and creativity over when, where, and how often you sequence with MinION and MinION Mk1C — portable and affordable, real-time sequencing devices.
Flow cells
Consumable components for MinION and MinION Mk1C devices that process the prepared DNA/RNA sample.
DNA library prep kits
Streamlined library preparation kits for nanopore sequencing, optimised for a wide range of applications — from direct sequencing of whole genomes or metagenomes to targeted sequencing of specific regions of interest.
RNA library prep kits
Sequence full-length transcripts and RNA viruses in single reads using nanopore technology. Use cDNA-based approaches for low-input amounts or directly sequence native RNA to include epigenetic modifications.
Expansion packs
Additional reagents and consumables to support your nanopore sequencing assays.
Spotlight: Real-time microbial sequencing
From rapid species identification and antimicrobial resistance profiling to generating complete genome assemblies — get immediate access to results with real-time, portable, and affordable MinION devices.