VWR Glassware
VWR borosilicate 3.3 glassware is characterized by a high degree of chemical resistance to water, acids, saline solutions, alkaline solutions and organic substances. Only hydrofluoric acid, strong alkaline solutions at higher temperature or hot, concentrated phosphoric acid can cause surface removal of the glass.