VWR Volumteric Flask
Avail special pricing
  • Buy 1.5 L worth volumetric flask @ 45% + 9% discount.
  • Buy 1.5 L worth VWR TRUE bottles @special price.
  • Get a surprise gift on purchase of 50,000 INR.
VWR glassware is produced from high quality borosilicate 3.3 glass which is due to its composition is characterized by excellent chemical resistance, high temperature resistance and very good resistance against thermal shock. Produced according to the international standard DIN ISO 3585 borosilicate 3.3 glass is therefore the ideal glass for mostly all laboratory applications. Chemical composition of VWR borosilicate 3.3 glass:
  • AS per highest quality of ISO/USP
  • Individually calibrated
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion;
Mel Articlenumber Material Description List
price
Offer Pricing
VWRA612-6829 Volumetric flask, 1ml, class A, ISO 6042 Buy Now
VWRA612-6830 Volumetric flask, 2ml, Class A, ISO 6042 Buy Now
VWRA612-6831 Volumetric flask 5ml, Class A, ISO 1815 Buy Now
VWRA612-6832 Volumetric flask 10ml, class A ISO 1577 Buy Now
VWRA612-6833 Volumetric flask 20ml, Class A ISO 1577 Buy Now
VWRA612-6834 Volumetric flask 25ml, Class A ISO 1454 Buy Now
VWRA612-6835 Volumetric flask 50ml, Class A, ISO 1378 Buy Now
VWRA612-6836 Volumetric flask 100ml, Class A, ISO 1340 Buy Now
VWRA612-6837 Volumetric flask 200ml, Class A, ISO 1900 Buy Now
VWRA612-6838 Volumetric flask 250ml, Class A, ISO 1900 Buy Now
VWRA612-6839 Volumetric flask 500ml, Class A, ISO 2489 Buy Now
VWRA612-6840 Volumetric flask 1000ml, Class A, ISO 3639 Buy Now
VWRA612-6841 Volumetric flask 2000ML, Class A, ISO 2955 Buy Now
VWRA612-6842 Volumetric flask 5ml, Class A, USP 2033 Buy Now
VWRA612-6843 Volumetric flask 10ml, Class A, USP 1767 Buy Now
VWRA612-6844 Volumetric flask 25Mml, Class A USP 1625 Buy Now
VWRA612-6845 Volumetric flask 50ml, Class A, USP 1549 Buy Now
VWRA612-6846 Volumetric flask 100ML, Class A, USP 1520 Buy Now
VWRA612-6847 Volumetric flask 200ML, Class A, USP 2128 Buy Now
VWRA612-6848 Volumetric flask 250ml, Class A, USP 2128 Buy Now
VWRA612-6849 Volumetric flask 500ML, Class A, USP 2812 Buy Now
VWRA612-6850 Volumetric flask 1000ml, Class A, USP 4085 Buy Now
VWRA612-6851 Volumetric flask 2000ml, Class A, USP 3320 Buy Now