VWR True Bottles
Avail special pricing
- Buy 1.5 L worth volumetric flask @ 45% + 9% discount.
- Buy 1.5 L worth VWR TRUE bottles @special price.
- Get a surprise gift on purchase of 50,000 INR.
VWR glassware is produced from high quality borosilicate 3.3 glass which is due to its composition is characterized by excellent chemical resistance, high temperature resistance and very good resistance against thermal shock. Produced according to the international standard DIN ISO 3585 borosilicate 3.3 glass is therefore the ideal glass for mostly all laboratory applications. Chemical composition of VWR borosilicate 3.3 glass:
- AS per highest quality of ISO/USP
- Individually calibrated
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion;
Mel Articlenumber | Material Description | List price |
Offer Pricing |
---|---|---|---|
VWRA215-4436 | Bottle reagent clear 25 ml with S. cap |
|
Buy Now |
VWRA215-4437 | Bottle reagent clear 50 ml with S. cap |
|
Buy Now |
VWRA215-4438 | Bottle reagent clear 100 ml with S. cap |
|
Buy Now |
VWRA215-4439 | Bottle reagent clear 250 ml with S. cap |
|
Buy Now |
VWRA215-4440 | Bottle reagent clear 500 ml with S. cap |
|
Buy Now |
VWRA215-4441 | Bottle reagent clear 1L with S. cap |
|
Buy Now |
VWRA215-4442 | Bottle reagent clear 2L with S. cap |
|
Buy Now |
VWRA215-4443 | Bottle reagent clear 5L with S. cap |
|
Buy Now |
VWRA215-4444 | Bottle reagent clear 10L with S. cap |
|
Buy Now |
VWRA215-4445 | Bottle reagent clear 20L with S. cap |
|
Buy Now |