VWR Glassware- "ARAMBH"
Avail special pricing
- Buy 1.5 L worth volumetric flask @ 45% + 9% discount.
- Buy 1.5 L worth VWR TRUE bottles @special price.
- Get a surprise gift on purchase of 50,000 INR.
VWR glassware is produced from high quality borosilicate 3.3 glass which is due to its composition is characterized by excellent chemical resistance, high temperature resistance and very good resistance against thermal shock. Produced according to the international standard DIN ISO 3585 borosilicate 3.3 glass is therefore the ideal glass for mostly all laboratory applications. Chemical composition of VWR borosilicate 3.3 glass:
- AS per highest quality of ISO/USP
- Individually calibrated
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion;