Masterflex® B/T® Series Peristaltic Pumps
Bulk transfer at its best
Highest flow rate
Flow range: 0.3 to 42 LPM
Makes the job efficient
Masterflex® B/T Series peristaltic pumps have the highest flow rates of any Masterflex® pump. Their benefits include:
- Peristaltic pump bulk transfer at its best
- Highest flow rate
- Flow ranges from 0.3 to 42 LPM
Our B/T Series pumps simply make your job more efficient. And with their heavy-duty motors and excellent hazard protection, they’re built to last.
Masterflex B/T Series Pump Components
Reimage fluid handling
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
A broad flow range, from 0.65 to 42 LPM (0.17 to 11.1 GPM), for integration into your higher-flow bioprocess applications. Pump head accepts B/T PerfectPosition® pump tubing sizes B/T 87 and B/T 91 for repeatable and reliable performance. Open-head sensor technology stops the drive motor when pump head is opened.
Custom services and tools
Custom engineered solutions
Partnering on custom engineered pumps – making ideas reality for over 50 years.
Learn more
Custom 'cut to length' tubing
We have the flexibility to meet your needs and save you time and money. Choose your tubing type, length and quantity.
Learn more
MasterflexLive®
With MasterflexLive®, you get real-time monitoring of all pump parameters.
Learn more
Masterflex Peristaltic Pump Series — Shop by flow range
Ismatec® Series
Microflow, high accuracy
Flow Range:
0.0001 to 365 mL/min
Microflow pumps >
L/S® Series
Precise flow control
Flow Range:
0.00005 to 3400 mL/min
Precision flow >
I/P® Series
Powerful systems
Flow Range:
0.001 to 26 LPM
Industrial flow >
B/T® Series
Highest flow rates
Flow Range:
0.3 to 42 LPM
Highest flow pumps >
Need more information or help selecting the perfect product for your application?
Our experts are ready to help.