Masterflex® MasterSense™ Gear & Piston Process Pumps
Your process, evolved
Using MasterSense™ digital technology, these pumps offer advanced user experience, sensor monitoring, flow control, and connectivity for mid to high flow, higher pressure applications.
Features that make your process effortless
Perfect for sanitary environments —
316 polished stainless steel housing IP66 NEMA 4X rated for washdown
Sensors integrated into the flow path monitor fluid pressure and alert before there is a potential for process failure.
Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity-
even with wet gloves on!
Over-the-air firmware updates ensure optimization of your pump system without moving the system
Superior mechanical design supports higher flow ranges, pressures and torque capability for greater repeatability and pumping higher viscosity fluids.
Gear Pumps are the optimal choice for pumping clean fluids in higher pressure, pulseless flow applications.
VIEW FULL SPECS
Piston Pumps offer high repeatability for critical dispense applications where change-out and lot-to-lot contamination is not critical.
VIEW FULL SPECS
Continous Mode*
Time Mode
Volume Dispense
Pressure
User Access
Program Save & Recall
Network Detection
Remote Mode*
Head Calibration*
Skip reading the operation manual
Intuitive touch screen interface, even when wearing gloves
- Easy to set up and program
- 5" capacitive TFT/RGB touch screen with a clear, wide viewing angle
- Adjust parameters with a simple touch - even with gloves on
- Operational modes include continuous run, volume dispense, and time dispense
- Batch control and interval/off time in Volume and Time modes
- Easily save and recall up to 50 commonly used protocols, each with a unique name
- Know when you hit your mark - graphical indicator lets you know when pump system is optimally calibrated
Integrated sensor monitors your media so you don’t have to
Seamless monitoring, avoid risk and save money
- Install in-line pressure sensor
- Connect sensor to pump
- Configure alarms and start pump
Advanced Connectivity and Control Options
Monitor from anywhere
Flexible Connectivity
- Your choice of connectivity options- EtherNet/IP™, PROFIBUS, Serial, or analog control
- Three-tier access to pump functionality—use this option to protect your process by controlling who can operate the pump and at what level
- Over-the-air firmware updates - automatically allows you to take full advantage of future enhancements
- Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
Gear pump complete system components
To create a complete pump system, a drive and pump head are needed. Complete systems are available for common configurations (see table below), or choose the pump drive and select from a variety of Suction shoe mount A or B, or cavity style head pump head options available.
Gear Pump Complete Specifications & Product Details |
||||
![]() |
![]() |
|||
Drive Only | Gear Pump System | |||
Model | MFLX07576-50 | MFLX77976-50 | MFLX77976-51 | MFLX77976-52 |
Flow range | 1.7 to 7380 ml/min | 9.2 to 552 ml/min | 64 to 3840 ml/min | 117 to 7000 ml/min |
RPM | 100 to 6000 RPM | |||
Max Differential Pressure | 8.6 bar (125 psi) | 4 bar (60 psi) | 6.9 bar (100 psi) | 1.7 (25 psi) |
Included pump Head | Order Separately | MFLX07002-14 | MFLX74012-21 | MFLX07003-04 |
Process connection | 1/8" NPT Female | 1/8" NPT Female | 1/8" NPT Female | |
Viscosity | 0.5 to 1500 Centipoise (Clear fluids) |
Learn more - See Gear pump performance and technical spec sheet
Gear pump heads
Piston pump complete system components
To create a complete pump system, a drive and pump head are needed. Complete systems are available for common configurations (see table below), or choose the pump drive and select from a variety of Q-series or Rh-Series Piston pump heads. An adapter required for low-flow RH models.
Gear Pump Complete Specifications & Product Details |
||||
![]() |
![]() |
|||
Drive Only | Gear Pump System | |||
Model | MFLX07576-50 | MFLX77976-50 | MFLX77976-51 | MFLX77976-52 |
Flow range | 1.7 to 7380 ml/min | 9.2 to 552 ml/min | 64 to 3840 ml/min | 117 to 7000 ml/min |
RPM | 100 to 6000 RPM | |||
Max Differential Pressure | 8.6 bar (125 psi) | 4 bar (60 psi) | 6.9 bar (100 psi) | 1.7 (25 psi) |
Included pump Head | Order Separately | MFLX07002-14 | MFLX74012-21 | MFLX07003-04 |
Process connection | 1/8" NPT Female | 1/8" NPT Female | 1/8" NPT Female | |
Viscosity | 0.5 to 1500 Centipoise (Clear fluids) |