5 Part Analyzers and Advance Analyzers
BeneSphera™ (A Brand of Avantor) - 3 Part Hematology Analyzers
Elevate your diagnostic capabilities with BeneSphera™ 3-Part Hematology Analyzers. Engineered for accuracy and efficiency, our cutting-edge analyzers provide a comprehensive assessment of blood components, enabling healthcare professionals to make informed decisions for optimal patient care. Explore the power of precision in hematology diagnostics with BeneSphera™.
Three-part hematology analyzers offer several advantages in clinical laboratory settings, providing a comprehensive yet efficient analysis of blood components. Here are some key advantages of 3-Part Analyzers,
- Cost-Effectiveness
- Efficiency in Routine Hematology Testing
- Quick Turnaround Time
- User-Friendly Operation
- Quality Control and Calibration
- Cost-Effective Consumables
BC 5150 5-Part Differential Hematology Analyzer
The BC 5150 Auto Hematology Analyzer is a light and compact system, specially tailored to assist diagnostic labs who need full CBC+5 part results, with a relatively low daily sample volume. Responding to increased demands for high quality read-outs and accurate diagnostic decisions, the BC 5150 provides a reliable complete 5-part differential menu, 25 reportable parameters and 12 research parameters, 3 histograms and 3 scattergrams.
See the product
BC 6200 Advanced Automated Hematology Analyzer
With the newly designed optics and reagent systems, the SF Cube technology can help doctors to better differentiate the clusters of cells, which is the key to revealing more abnormal cells
See the Product