3 Part Analyzers
BeneSphera™ (A Brand of Avantor) - 3 Part Hematology Analyzers
Elevate your diagnostic capabilities with BeneSphera™ 3-Part Hematology Analyzers. Engineered for accuracy and efficiency, our cutting-edge analyzers provide a comprehensive assessment of blood components, enabling healthcare professionals to make informed decisions for optimal patient care. Explore the power of precision in hematology diagnostics with BeneSphera™.
Three-part hematology analyzers offer several advantages in clinical laboratory settings, providing a comprehensive yet efficient analysis of blood components. Here are some key advantages of 3-Part Analyzers,
- Cost-Effectiveness
- Efficiency in Routine Hematology Testing
- Quick Turnaround Time
- User-Friendly Operation
- Quality Control and Calibration
- Cost-Effective Consumables
BENESPHERA™ H33S HEMATOLOGY ANALYSER
CL-1000i is a robust and simple-to-use chemiluminescence analyzer with a throughput of up to 120 tests per hour. In a compact benchtop package, it has a large onboard capacity of 25 reagents and 60 samples, and supports non-stop refill of samples, reagents and consumables during testing. By integrating the reliable CLIA technology with latest electrical and software improvement, CL-1000i enables easy operation and brings confident diagnosis faster than ever.
See the product
BENESPHERA™ H 31 PLUS HEMATOLOGY ANALYSER
The BeneSphera™ H33s is a compact, powerful, full featured and affordable system. It provides a detailed flag information never before seen on a 3-part analyzer. The system provides information useful for diagnosis including WBC flag, RBC flag and PLT flag.
See the product