Electrolytes
BeneSphera™ (A Brand of Avantor) - Electrolyte Product Range
Elevating Electrolyte Diagnostics to New Heights
BeneSphera is committed to advancing electrolyte diagnostics to improve patient outcomes. Our Electrolyte Product Range reflects this dedication, providing laboratories with the tools they need to deliver precise and reliable electrolyte results.
BeneSphera™ Electrolyte Analyzer - (Na / K / iCa / Li / pH/Cl)
Automated analyzer for measurement of Na / K / iCa / Li / pH/Cl in serum, plasma, whole blood, diluted urine and cerebrospinal fluid.
See the product