THREE in a ROW- VWR PHP Pipettes
Get 3 for the price of 1
- Buy 3 PHP pipettes @ 1 pipette's price
- Get 3 years of warranty with each PHP Pipettes
- Buy 3 pipettes of any volume and get pipette stand free
VWR PHP Pipettes are Precise High Performance, fully autoclavable pipettes, that are available in Single and Multi Channel range. VWR PHP offers one of the lowest plunger forces and a durable tip cone. The tip ejector is made using special corrosion resistant alloy, protecting it against chemical and physical corrosion. The pipette has an effortless tip ejection with unique shock absorbing mechanism. The tip cone is spring loaded for lesser strain and proper tip fitment for precise pipetting (for Multi Channel only).
- Superior accuracy & precision (tested in ISO 17025 accredited lab)
- Magnet assisted piston for low plunger force and lesser chance of RSI
- PVDF tip cone for durable performance
- Large and clear 4-digit display; color coding for easy identification
- 3-Year warranty
Mel Articlenumber | Material Description | List Price | 3 in 1 Pricing |
---|---|---|---|
VWRA613-6710 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 100 |
|
Buy Now |
VWRA613-6709 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 500 |
|
Buy Now |
VWRA613-6708 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 200 |
|
Buy Now |
VWRA613-6707 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 100 |
|
Buy Now |
VWRA613-6706 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 500 |
|
Buy Now |
VWRA613-6705 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 250 |
|
Buy Now |
VWRA613-6704 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 200 |
|
Buy Now |
VWRA613-6703 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 100 |
|
Buy Now |
VWRA613-6702 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 50 |
|
Buy Now |
VWRA613-6701 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 25 |
|
Buy Now |
VWRA613-6700 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 20 |
|
Buy Now |
VWRA613-6699 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 10 |
|
Buy Now |
VWRA613-6698 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 5 |
|
Buy Now |
VWRA613-6697 | PHP - SINGLE CHANNEL FIX VOL PIPETTE 2.5 |
|
Buy Now |
VWRA613-6696 | PHP - SINGLE CHANNEL VV PIPETTE 1000-10000 |
|
Buy Now |
VWRA613-6695 | PHP - SINGLE CHANNEL VV PIPETTE 1000-5000 |
|
Buy Now |
VWRA613-6694 | PHP - SINGLE CHANNEL VV PIPETTE 100-1000 |
|
Buy Now |
VWRA613-6693 | PHP - SINGLE CHANNEL VV PIPETTE 20-200 |
|
Buy Now |
VWRA613-6692 | PHP - SINGLE CHANNEL VV PIPETTE 10-100 |
|
Buy Now |
VWRA613-6691 | PHP - SINGLE CHANNEL VV PIPETTE 5-50 |
|
Buy Now |
VWRA613-6690 | PHP - SINGLE CHANNEL VV PIPETTE 2-20 |
|
Buy Now |
VWRA613-6689 | PHP - SINGLE CHANNEL VV PIPETTE 0.5-10 |
|
Buy Now |
VWRA613-6688 | PHP - SINGLE CHANNEL VV PIPETTE 0.1-2 |
|
Buy Now |