BAKERBOND Q2100 columns – “Quality for the 21st century”
Avantor® manufacture Bakerbond Q2100 UHPLC/HPLC column are based on ultra-high purity silica. Available with 1.8 µm, 3 µm, 5 µm Particle sizes with narrow particle size distribution. Bakerbond phases achieve high efficiency separations with available four different chemistries (C18, C8, Phenyl-Hexyl and Silica)
Bakerbond Q2100 LC phases have specially designed to solve the challenges of most of the routine samples as well in new method development. With four widely used chemistries (C18, C8, Phenyl-Hexyl and Silica) and three commonly used particle sizes (1.8 µm, 3 µm, 5 µm ) Bakerbond can be an useful alternative to any of the regular column you use.
Contact Technical support
Download Bakerbond brochure
BAKERBOND Q2100 columns – “Quality for the 21st century”
  • High efficiency UHPLC/HPLC columns
  • C18, C8, Phenyl-Hexyl and Silica Phases
  • Available with 1.8 µm, 3 µm, 5 µm particle sizes
Phase Guide
See the products