ELISA
BeneSphera™ (A Brand of Avantor) - ELISA ( Reagent & Instruments)
Unleashing Precision in Diagnostics.
Explore our comprehensive suite of enzyme-linked immunosorbent assay (ELISA) products, meticulously crafted for accurate and reliable results across diverse clinical applications.
Enzyme-Linked Immunosorbent Assay (ELISA) plays a crucial role in the field of diagnostics and research, offering several key advantages that contribute to its widespread importance. Here are some aspects highlighting the significance of ELISA:
- High Sensitivity and Specificity
- Versatility in Applications
- Quantitative and Qualitative Analysis
- Cost-Effectiveness
- Widespread Availability of Reagents and Kits
- Long-Term Stability of Results
BeneSphera™ HIV Advanced Elisa Kit
BeneSphera™ HIV Advanced Elisa Kit is a fourth-generation immunoassay for qualitative detection of P24 antigen and antibodies to HIV-1 and HIV-2.
See the product
BeneSphera™ HbsAg Elisa Kit
BeneSphera™ HbsAg Elisa Kit is for qualitative detection of for detection Hepatitis B Surface Antigen from human serum & plasma
See the product
BeneSphera™ HCV Elisa Kit
BeneSphera™ HCV Elisa Kit is for qualitative detection of antibodies to HCV from human serum & plasma
See the product
BeneSphera™ T3 Elisa Kit
BeneSphera™ T3 Elisa Kit is for quantitative determination of T3 (Triiodothyronine) Hormone from human serum & Plasma
See the product
BeneSphera™ T4 Elisa Kit
BeneSphera™ T4 Elisa Kit is for quantitative determination of T4 (thyroxine) Hormone from human serum & Plasma
See the product
BeneSphera™ TSH Elisa Kit
BeneSphera™ TSH Elisa Kit is for quantitative determination of TSH (Thyroid Stimulating Hormone ) Hormone from human serum & Plasma
See the product
BeneSphera™ FT3 Elisa Kit
BeneSphera™ FT3 Elisa Kit is for quantitative determination of FT3 (Free Triiodothyronine) Hormone from human serum & Plasma
See the product
BeneSphera™ FT4 Elisa Kit
BeneSphera™ fT4 Elisa Kit is for quantitative determination of FT4 (Free thyroxine) Hormone from human serum & Plasma
See the product
BeneSphera™ FSH Elisa Kit
BeneSphera™ FSH Elisa Kit is for quantitative determination of FSH (Follicle-stimulating hormone) Hormone from human serum & Plasma
See the product
BeneSphera™ Prolactin Elisa Kit
BeneSphera™ Prolactin Elisa Kit is for quantitative determination of Prolactin Hormone from human serum & Plasma
See the product
BeneSphera™ LH Elisa Kit
BeneSphera™ LH Elisa Kit is for quantitative determination of LH (Luteinizing hormone) Hormone from human serum & Plasma
See the Product