Glove box, fibreglass, HEPA filtered, Protector®
Supplier: LABCONCO
5065035 5065031
LBCP5065035EA 0 INR
LBCP5065035 LBCP5065031
Glove box, fibreglass, HEPA filtered, Protector®
Glove Boxes and Glove Bags
This filtered glove box with fibreglass liner provides inlet and outlet HEPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials. The glove box may also be appropriate for use as a containment isolator for applications that include compounds assigned ISO 4 and ISO 5 Banding Levels.
  • LCD display provides real time monitoring of volume dilution rates in cubic feet per minute (CFM), air changes/minute (ACM), and main chamber pressure (inches of water)
  • Built-in blower with speed control capable
More Product Information