Laboratory fume hoods, benchtop, Protector® XL
Supplier: LABCONCO
111820021 111510020
LBCP111820021EA 1421360 INR
LBCP111820021 LBCP111510020
Laboratory fume hoods, benchtop, Protector® XL
Fume Hoods and Enclosures Chemical Fume Hoods
The Protector® XL benchtop laboratory hoods offer superior design flexibility. Their panel liners are supplied in three depths to meet individual installation and application requirements. These are suitable for general chemistry applications.
  • By-pass airflow design
  • Glacier white powder-coated steel exterior
  • Chemical-resistant molded composite panel liner and pre-set baffle(s) with flame spread less than 25 per ASTM E-84
  • Tempered safety glass vertical-rising sash(es) with powder-coated aluminum frame(s)
  • Curved air foil allows air to sweep the work surface
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable