Shaking Water Baths, SW22 and SW23
Supplier: JULABO GMBH
9550322 9550323
462-4102EA 0 INR
462-4102 462-4104
Shaking Water Baths, SW22 and SW23
Baths Shaking Water Baths
Stainless steel shaking water baths are suitable for unsupervised continuous operation. The bright, LED Multi-Display showing up to six temperature values is visible from across the lab. Advanced microprocessor technology with PID temperature control eliminates temperature overshoot and provides high temperature stability. Sensitive samples are well protected; as soon as the high and low temperature limits are reached an intermittent tone sounds. If the liquid in the bath reaches a minimum, the unit shuts down completely and a continuous single tone sounds.
  • Electronic timer for setting
More Product Information