Bath fluids
Supplier: JULABO GmbH
8940115 8940101 8940103 8940104 8940105 8940107 8940109 8940102 8940133 8940132 8940141
461-0226EA 0 INR
461-0226 461-0110 461-0120 461-0131 461-0130 461-0140 461-0225 461-0121 461-0478 461-0477 461-0878
Bath fluids
Circulator Bath Fluids
Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.
  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness