Centrifugal concentrators, complete systems and system packages, CentriVap®
Supplier: LABCONCO
7982011 7983014 7989601 7315032 7315030 7315033 7315031
531-0223EA 0 INR
531-0223 531-0224 535-0409 431-0235 431-0231 431-0237 431-0233
Centrifugal concentrators, complete systems and system packages, CentriVap®
Evaporators Centrifugal Evaporators
CentriVap® vacuum concentrators are available as complete systems or system packages. The acid resistant model is recommended for corrosive solvents (TFA, acetic acid, and so on). Other models feature Heat Boost, which surrounds the chamber wall, to further speed evaporation of high boiling point solvents.
  • Epoxy-coated aluminium chamber
  • Centrifugation, controlled by microprocessor, up to 1725 min⁻¹ and heating, controlled by microprocessor up to 100 °C
  • Control panel with on/off button, pre-heating button, optional Heat Boost button, parameter setting,
More Product Information