Upright ULT Freezers, -40 to -86 ℃
Avantor VWR's new range of Ultra-Low Freezers will fit all your long cold storage needs.
The VWR Ultra-low temperature freezers are designed for safe and reliable storage of sensitive samples. Our wide range of freezers offer a host of benefits:
Ergonomic design for easy door opening and closing.Lockable with ability to add a padlock for extra security
Microprocessor controlled with user settable parameters such as set point and alarms
Standard USB port capable of storing >15 years of operating data for compliance
VIP insulation and multilayered sealing design with 70mm insulating layer to improve energy efficiency and reduce heat loss for excellent warm up times in event of power failure
Tool free removable filter design,easy and quick to clean the filter
4 casters +2-foot locks easy to move,lock and level
Choice
Enough to cover almost every application packed into a series of models with a wide selection of accessories
High Performance
Market-leading consistency and reliability backed up with warranties that run into many years
Care-free sample protection
Alarm systems with built-in audible/visual warnings for high/low temperature, door ajar, or power failure alert
Superior energy savings
Microprocessor-controlled system designed for a controllable range of -40 to -86 °C for cabinet space with 1 °C increments
SPECIFICATIONS
Cat. No. | VWRA471-1274 | VWRA471-1275 | VWRA471-1276 | VWRA471-1277 | VWRA471-1279 | VWRA471-1282 |
Defrost | Manual | Manual | Manual | Manual | Manual | Manual |
Alarm | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms | High/low temperature, hot condenser, power failure, high/low voltage, sensor error, low battery, high ambient temperature and door ajar alarms |
Electrical | 220-240 V, 50 Hz, 3 A | 220-240 V, 50 Hz, 7.5 A | 220-240 V, 50 Hz, 8 A | 220-240 V, 50 Hz, 8 A | 220-240 V, 50 Hz, 10 A | 220-240 V, 50 Hz, 10 A |
Noise level | 46.8 dB(A) | 50 dB(A) | 50 dB(A) | 50 dB(A) | 50 dB(A) | 50 dB(A) |
Shelves/drawers | 1 | 3 | 3 | 3 | 3 | 3 |
Door type | Single door, 2 inner doors | Single door, 2 inner doors | Double door, 4 inner doors | Single door, 4 inner doors | Single door, 4 inner doors | Single door, 4 inner doors |
Access ports | 1 | 2 | 2 | 2 | 2 | 2 |
Weight | 108 kg | 255 kg | 295 kg | 300 kg | 345 kg | 380 kg |