Photometry
Featured Solutions
The SpectraMax® QuickDrop Micro-Volume Spectrophotometer addresses the needs of life science researchers who quantify very small amounts of DNA, RNA, oligos, proteins and other compounds and allows you to set up your laboratory with a minimal investment of time, cost, and effort.
mySPEC allows you to perform nucleic acid and proteins microvolume concentration plus purity measurements.