Instrument Systems & Modules
VWR International provides combinations of hardware and software along with a comprehensive range of accessories and specialised technical services – in fact a complete solution for your analytical or preparative HPLC applications in one area.
The LaChromUltra™ system, in combination with ultra fast columns packed with particles of ≤2 μm, enable conventional HPLC methods to be carried out up to 10 times quicker giving you an amazing increase in productivity.
- LaChromUltra™
- LaChrom Elite®
- LC-MS systems
- System for post-column derivatisation
- Amino acid analyser
Download Chromaster Ultra RS brochure