Respiration BOD: BOD self-checks
A reduction in oxygen (consumed by micro organisms) causes a definite pressure difference that is measured by a pressure sensor. Practical and easy to perform!
Applications for BOD respirometric methods
BOD, OxiTop®, WTW
Respirometric OxiTop® measuring system for BOD(5) self-check measurement with undiluted samples (up to 4000 mg/l BOD).
More Information