Cleanroom Mop Systems
For easy cleaning of standard equipment in cleanrooms and controlled environments. The hardware is constructed to deal with the space restrictions of isolators or hoods and to reach small pass thrus, to clean 'over and under' (with collapsible handles made from electropolished stainless steel), as well as to clean around and under heavy process equipment or calibrated apparatus.
Curtain cleaners are designed with hard wearing, low linting slip covers (a selection of different slipcovers is available) for an easy cleaning of curtains and softwall in cleanrooms and in critical and sterile environments.
Product selection
Mop Systems
Please choose from a list of products and refine your search by attributes...