SNAP i.d.® 2.0 Protein Detection Systems IHC
Supplier: MILLIPORE
SNAP2IHC2 SNAP2IHC
MILYSNAP2IHC2EA 0 INR
MILYSNAP2IHC2 MILYSNAP2IHC
SNAP i.d.® 2.0 Protein Detection Systems IHC
Analysers Protein Analysers
The SNAP i.d.® 2.0 System for IHC introduces a streamlined, medium through-put IHC capability to our SNAP i.d. 2.0 family.
  • Eliminates the need for pap pens
  • Antibodies can be collected and reused
  • Slide handling time is significantly decreased Less time spent on wash steps
  • Parallel processing of multiple slides
  • Flexibility of multiple slide configurations enables the processing of 1 to 24 slides at a time
  • Compatible with standard IHC slides and protocols
  • Compatible with diverse tissue preparations including formalin-fixed or fresh frozen samples
For use with immunohistochemistry slides.
Delivery information:
This system includes the SNAP i.d. 2.0™ Base, IHC frame and incubation cover, assembly fixture, vacuum tubing, and a quick start guide.