Refrigerators, wine storage cabinets
Supplier: Liebherr
WK4127
471-0377EA 0 INR
471-0377
Refrigerators, wine storage cabinets
Refrigerators
Cabinets designed for storage and ageing of wines with a protected environment.
  • Digital temperature display
  • Height-adjustable wooden grid shelves
  • High-quality SwingLine design, reversible doors and subsequent lock
  • Forced air cooling and fresh air supply via activated charcoal filter
  • Vibration-free compressors