tri-Sodium citrate dihydrate

Supplier: BDH (US PRODUCED)

Synonyms: Sodium Citrate, DihydrateCitric acid trisodium salt dihydrateSodium Citrate DihydrateSodium citrate tribasic dihydrateSodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate

BDHB0288-2.5KGEA 0 INR
BDHB0288-2.5KG
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294,1 g/mol
Melting Pt: 150 °C
Density: 1,76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://in.vwr.com/stibo/web/std.lang.all/06/03/16640603.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR