tert-Butanol ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol
9056-01EA
15074.87
INR
9056-01
9056-05
tert-Butanol ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
tert-Butanol
Formula:
(CH₃)₃COH MW: 74.12 g/mol Boiling Pt: 82.2 Melting Pt: 25.3 Density: 0.7887 g/cm³ (20 °C) Flash Pt: 11 (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004464 CAS Number: 75-65-0 EINECS: 200-889-7 UN: 1120 ADR: 3 II Merck Index: 12,01577 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | - |
For Laboratory,Research,or Manufacturing Use |
|
NOTE: Product freezes at about 25°C (77°F). |
|
ACS - Assay ((CH₃)₃COH) (by GC, corrected for water) | ≥ 99.0 % |
Carbonyl Compounds - Formaldehyde | ≤ 0.01 % |
Color (APHA) | ≤ 20 |
Residue after Evaporation | ≤ 0.003 % |
Titrable Acid (meq/g) | ≤ 0.001 |
Water (H₂O)(by Karl Fischer titrn) | ≤ 0.1 % |
Product Information (not specifications): | 81.5 - 83.0 °C. |
Appearance (white, crystalline solid; colorless liquid above 25.5°C) | 0.781- 0.785 |
Boiling Range = 81.5 - 83.0 °C. | 24 °C. |
Specific Gravity at 25°/25°C = 0.781- 0.785 | |
Freezing Point (typical) = 24 °C. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...