Bacitracin
Supplier: PanReac AppliChem
A0623.0025EA
0
INR
A0623.0025
Bacitracin
Bacitracin
Formula:
C₆₆H₁₀₃N₁₇O₁₆S₁ MW: 1421,60 g/mol |
MDL Number:
MFCD00062640 CAS Number: 1405-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...