Hydrochloric acid 1 mol/l (1 N)
Supplier: BDH (US PRODUCED)
|
Warning
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
3202-7
3202-1
BDHB3202-7EA
0
INR
BDHB3202-7
BDHB3202-1
Hydrochloric acid 1 mol/l (1 N)
Hydrochloric acid
Formula:
HCl MW: 36,46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...