Magnetic Stirrers, Spherical
Supplier: COWIE
442-9801EA 0 INR
442-9801
Magnetic Stirrers, Spherical
PTFE, specifically designed for use in tubes and eccentric stirring.
  • Good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (-200 to +250 °C)