VWR® Saf-T Tainer® Biomedical Waste Containers
Supplier: Avantor
VWR® Saf-T Tainer®
119-0907EA 0 INR
119-0907
VWR® Saf-T Tainer® Biomedical Waste Containers
Bins Waste Bins
Sharps container systems provide solutions for the safe and effective containment and disposal of contaminated needles, syringes and other sharps.
  • Three-component biomedical waste containers
  • Puncture- and leak-resistant
  • Incinerator-safe
  • Systems store flat, and feature locking lids and an integrated handle
These three-component biomedical waste containers consist of an outer corrugated container and an inner corrugated container separated by a red plastic liner.