VWR® Advanced 700, Magnetic Hotplate Stirrer
Supplier: VWR Collection
444-2862EA 0 INR
444-2862 444-2866 444-2868 444-2870
VWR® Advanced 700, Magnetic Hotplate Stirrer
Stirrers Magnetic Hotplate Stirrers
VWR® 700 Series Advanced hotplate stirrers deliver accurate and repeatable results. Low profile, space saving design takes up less space in the laboratory. Spill-resistant housing channels fluids away from internal components. Front panel features easy to use controls, which allow users to dial-in adjustments. Integral support rod holder with locking knob accepts the optional support rod and clamp kit. Microprocessor control regulates heating and stirring functions. Samples are heated to temperature quickly and accurately. Speed is precisely controlled, giving constant stirring throughout the entire range. Ramping feature slowly increases speed to avoid splashing, improves magnetic coupling and provides excellent low speed control.
  • LED display and microprocessor control
  • Over-temperature and stir protection
  • On-board memory recalls last set temperature
  • Ceramic or aluminium top plate
  • Hot warning when plate is in use
Ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean. Durable aluminium tops will not crack or chip, and provide a more even heating surface. For additional safety, a hot symbol warning light is illuminated when heat is turned on and remains on until top plate cools down. Stir protection automatically shuts down the heater if the motor stops, over temperature protection ensures unit will shut down if it exceeds the set temperature by 10 °C. Built-in memory allows users to recall last set temperature even when unit has been turned off.
Delivery information: Each unit is supplied complete with 2 plugs: EU and UK, and a PTFE coated magnetic stirring bar.
Caution: * Below 100 °C ±2 °C Environmental and sample conditions permitting.