Ethyl acetate 99,50 ACS, Analytical reagent, Rankem®

Supplier: RANKEM
Danger

Synonyms: Acetic acid ethyl ester

E0082 E0083 E0081
RANKE0082EA 2432.1 INR
RANKE0082 RANKE0083 RANKE0081
Ethyl acetate 99,50 ACS, Analytical reagent, Rankem®
Ethyl acetate

High purity analytical reagent that meets or exceeds the ACS reagent specifications.


  • Batch-to-batch consistency
  • Tested for heavy metals
  • Suitable for analytical testing
  • Available in various pack size


Description: clear, colourless liquid.

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: −4 °C
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
Merck Index: 13,03792

Order Now

Specification Test Results

Color (APHA) ≤ 10
Guarantee Analysis .
Assay (GC) (CH3COOCH2CH3) ≥ 99.50 %
Weight Per ml (at 20°C) 0.899 - 0.901 g
Identity (IR) Conforms
Maximum Limits Of Impurities .
Residue After Evaporation ≤ 0.001 %
Titrable Acid ≤ 0.0009 meq/g
Water ( By KF) ≤ 0.05 %
Substances Darkened By Sulphuric Acid Passes test
Methanol (GC) ≤ 0.10 %
Methyl Acetate (GC) ≤ 0.10 %
Methyl Formate (GC) ≤ 0.10 %
Copper(Cu) ≤ 0.20 ppm
Iron (Fe) ≤ 0.10 ppm
Lead ( Pb) ≤ 0.10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR