Petroleum spirit, 60…80 °C, Laboratory Reagent, Rankem®

Supplier: RANKEM
Danger

Synonyms: Petroleum ether, Petroleum benzine

P0100 P0110 P0115 P0720
RANKP0100EA 622.55 INR
RANKP0100 RANKP0110 RANKP0115 RANKP0720
Petroleum spirit, 60…80 °C, Laboratory Reagent, Rankem®
Petroleum spirit (8032-32-4)

General lab reagent for routine laboratory use.


  • Suitable for routine laboratory usage and classical analysis.


A clear, colourless liquid.

Boiling Pt: 60…80 °C (1013 hPa)
Density: 0,67 g/cm³ (20 °C)
Flash Pt: –40 °C
MDL Number: MFCD00081849
CAS Number: 8032-32-4
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Guarantee Analysis
Weight Per ml (at 20 °C) 0.664 --> 0.671 g
Boiling Range (95% v/v) 60.0 - 80.0 °C
Color ≤ 10 APHA
Maximum Limits Of Impurities
Residue On Evaporation Max. 0.002%
Free Acid (As CH₃COOH) Max. 0.005%
Water (By KF) Max. 0.05%
Readily Carbonisable Substances Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR