Hydrochloric acid min 35,40%, Analytical reagent (AR), Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
H0080
H0250
H0070
RANKH0080EA
1616
INR
RANKH0080
RANKH0250
RANKH0070
Hydrochloric acid min 35,40%, Analytical reagent (AR), Rankem®
Hydrochloric acid
High purity analytical reagent.
- Batch-to-Batch consistency
- Tested for heavy metals
- Suitable for analytical testing
- Available in various pack size
A clear, colourless fuming liquid.
Formula:
HCl MW: 36.46 g/mol Density: 1.18 g/cm³ (20 °C) |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II |
Specification Test Results
Guarantee Analysis | |
Assay | Min. 35.40% |
Specific Gravity | 1.170 --> 1.190 |
Maximum Limits Of Impurities | |
Residue On Ignition (As SO₄²⁻) | Max. 0.00100% |
Free Chlorine | ≤ 0.0002% |
Sulphate (SO₄²⁻) | ≤ 0.0005% |
Sulphite | ≤ 0.0001% |
Ammonium (NH₄⁺) | ≤ 0.0003% |
Arsenic (As) | ≤ 0.000002% |
Iron (Fe) | ≤ 0.00004% |
Heavy Metal (As Pb) | ≤ 0.00005% |
Copper (Cu) | Max. 0.000010% |
Aluminium (Al) | Max. 0.000050% |
Phosphate (PO₄³⁻) | ≤ 0.00005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...