Petroleum spirit, 40…60 °C, Laboratory Reagent, Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Petroleum ether, Petroleum benzine
P0070
P0080
P0085
RANKP0070EA
952.88
INR
RANKP0070
RANKP0080
RANKP0085
Petroleum spirit, 40…60 °C, Laboratory Reagent, Rankem®
Petroleum spirit (8032-32-4)
General lab reagent for routine laboratory use
- Suitable for routine laboratory usage and classical analysis
- A clear, colourless liquid.
Boiling Pt:
40…60 °C (1013 hPa) Density: 0,645…0.665g/cm³ (20 °C) Flash Pt: –40 °C |
MDL Number:
MFCD00081849 CAS Number: 8032-32-4 EINECS: 265-151-9 UN: 1268 ADR: 3,II |
Specification Test Results
Guarantee Analysis | |
Weight Per ml (at 20 °C) | 0.640 --> 0.655 g |
Distillation Range (90% v/v) | 40.0 - 60.0 °C |
Maximum Limits Of Impurities | |
Residue On Evaporation | Max. 0.002% |
Free Acid (As CH₃COOH) | Max. 0.005% |
Water (By KF) | Max. 0.05% |
Readily Carbonisable Substances | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...