Petroleum spirit, 60…80 °C, Analytical reagent (AR), Rankem®

Supplier: RANKEM
Danger

P0084 P0090 P0092
RANKP0084EA 6388.38 INR
RANKP0084 RANKP0090 RANKP0092
Petroleum spirit, 60…80 °C, Analytical reagent (AR), Rankem®
Petroleum spirit (8032-32-4)

High purity analytical reagent


  • Batch-to-Batch consistency
  • Tested for heavy metals
  • Suitable for analytical testing
  • Available in various pack size


A clear, colourless liquidwith a characteristic odour

Boiling Pt: 60…80 °C (1013 hPa)
Density: 0,67 g/cm³ (20 °C)
Flash Pt: –40 °C
MDL Number: MFCD00081849
CAS Number: 8032-32-4
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Guarantee Analysis
Distillation Range 60.0 - 80.0 °C
Weight Per ml (at 20 °C) 0.664 --> 0.671 g
Maximum Limits Of Impurities
Non volatile matter Max. 0.001%
Sulphur Compounds (As SO₄²⁻) ≤ 0.0003%
Free Acid (As CH₃COOH) Max. 0.002
Copper (Cu) Max. 0.0001%
Iron (Fe) Max. 0.0001%
Lead (Pb) Max. 0.0001%
Water (By KF) Max. 0.02%
Bromine Number Max. 1.00


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR