Acetone ≥99.80%, Analytical reagent (AR), Rankem®
Supplier: RANKEM
|
Danger
|
A0136
A0137
A0138
RANKA0136EA
732.05
INR
RANKA0136
RANKA0137
RANKA0138
Acetone ≥99.80%, Analytical reagent (AR), Rankem®
Acetone
High purity analytical reagent that meets or exceeds the ACS reagent Specifications
- Batch-to-Batch consistency
- Tested for heavy metals
- Suitable for analytical testing
- Available in various pack size
Clear liquid and free from any suspended matter
No turbidity observed on mixing with distilled water at 27 ± 2 °C.
Formula:
CH₃COCH₃ Boiling Pt: 56,2 °C (1013 hPa) Melting Pt: –95,4 °C Density: 0,792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II |
Specification Test Results
BIS Specification | . |
Guarantee Analysis | . |
Purity | ≥ 99.70% |
Relative Density at 27/27 °C | 0.784 - 0.786 |
Color | ≤ 10 APHA |
Maximum Limits Of Impurities | . |
Residue On Evaporation, mg/100 ml | ≤ 5.00 |
Water content | ≤ 0.40% |
Acidity (As CH₃COOH) g/100 ml | ≤ 0.002 |
Alkalinity | Passes test |
Permanganate Test | ≥ 30 min |
Alcoholic Impurities | Passes test |
Benzene content | ≤ 2.00 mg/kg |
Methanol content | ≤ 300.0 mg/kg |
AR, ACS Specifications | |
Identity (IR) | Conforms |
Assay (GC) (CH₃)₂CO | ≥ 99.8 % |
Weight Per ml (at 20 °C) | 0.790 - 0.791 g |
Residue After Evaporation | ≤ 0.0005% |
Titrable Acid | ≤ 0.0003 meq/g |
Titrable Base | ≤ 0.0006 meq/g |
Water (By KF) | ≤ 0.20 % |
Aldehyde (As Hcho) | ≤ 0.002% |
Iso Propyl Alcohol (GC) | ≤ 0.05 % |
Copper (Cu) | ≤ 0.01 ppm |
Cobalt (Co) | ≤ 0.05 ppm |
Iron (Fe) | ≤ 0.10 ppm |
Lead (Pb) | ≤ 0.01 ppm |
Manganese (Mn) | ≤ 0.01 ppm |
Nickel (Ni) | ≤ 0.05 ppm |
Zinc (Zn) | ≤ 0.01 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...