Toluene ≥99.50%, Analytical reagent (AR), Rankem®

Supplier: RANKEM
Danger

T0096 T0097
RANKT0096EA 698.78 INR
RANKT0096 RANKT0097
Toluene ≥99.50%, Analytical reagent (AR), Rankem®
Toluene

High purity analytical reagent that meets or exceeds the ACS reagent Specifications


  • Batch-to-Batch consistency
  • Tested for heavy metals
  • Suitable for analytical testing
  • Available in various pack size


A clear, colourless liquid

Formula: C₆H₅CH₃
Boiling Pt: 110,6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0,867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 01-2119471310-51
Merck Index: 13,09607

Order Now

Specification Test Results

Color (APHA) ≤ 10
Guarantee Analysis
Assay (GC) (C₆H₅CH₃) ≥ 99.50%
Weight Per ml (at 20 °C) 0.865 - 0.870 g
Identity (IR) Conforms
Maximum Limits Of Impurities
Residue After Evaporation ≤ 0.001%
Titrable Acid ≤ 0.0002 meq/g
Titrable Base ≤ 0.0002 meq/g
Water (By KF) ≤ 0.03 %
Sulphur Compounds (As S) ≤ 0.003%
Thiophene Homologues % ≤ 0.0002%
Substances Darkened By Sulphuric Acid Passes test
Benzene (GC) ≤ 0.05 %
Cadmium (Cd) ≤ 0.01 ppm
Copper (Cu) ≤ 0.01 ppm
Iron (Fe) ≤ 0.05 ppm
Lead (Pb) ≤ 0.01 ppm
Zinc (Zn) ≤ 0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR