IL-6 Rapid Quantitative Test
Supplier: Finecare
FINCW251P0001EA 6250 INR
FINCW251P0001
IL-6 Rapid Quantitative Test
Assays Immunological Assays
Help identifying severe inflammatory response in patients with COVID-19. Act as an early alarm signal of 'cytokine storm' in COVID-19.
  • Aid in the management of critically ill patients
  • Results ready in 15 minutes
  • Room temperature storage up to 24 months
Early diagnosis of infection. Prognosis evaluation. Monitor the disease severity, evaluate after-surgery. IL-6 elevation can predict the worsening of 2019-nCoV.