Triethylamine ≥99.00%, Laboratory Reagent, Rankem®

Supplier: RANKEM
Danger

Synonyms: (Diethylamino)ethane, TEA

T0210 T0215
RANKT0210EA 955.9 INR
RANKT0210 RANKT0215
Triethylamine ≥99.00%, Laboratory Reagent, Rankem®
Triethylamine

General lab reagent for routine laboratory use .


  • Suitable for routine laboratory usage and classical analysis
  • Available in various pack size


Description: A clear colourless liquidwith a characteristic odour.

Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0,729 g/cm³ (20 °C)
Flash Pt: −11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 II
Merck Index: 13,09740

Order Now

Specification Test Results

Miscibility Miscible with ethanol andether.Miscible with waterbelow 19 deg.C. Passes test
Guarantee Analysis .
Assay (Gc) Min. 99.00%
Weight Per Ml (At 20 °C) 0.726 --> 0.728 g
Refractive Index (At 20 °C) 1.400 --> 1.403
Distillation Range 88.0 - 90.0 °C
Maximum Limits Of Impurities .
Water ( By Kf) Max. 0.200%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR