VWR® Sonic A Ultrasonic Bath
Supplier: Avantor
VWR®
142-0178EA 0 INR
142-0178 142-0179 142-0168 142-0169 142-0180 142-0181 142-0170 142-0171 142-0172 142-0173 142-0174 142-0175 142-0176 142-0177
VWR® Sonic A Ultrasonic Bath
Ultrasonic Baths
Ultrasonic cleaning bath with LED display, degas, sweep mode.
  • User-friendly control usability
  • Ultrasonic output stabilisation
  • Safety protection embedded microcontroller. Embedded microcontroller protection unit
The embedded/integrated microcontroller unit automatically controls and protects operating voltage, ultrasonic power output and heating temperature when the system is initialising, in operation and heating.
The shape of handles has been redesigned to provide aesthetic appearance and increased carrying capacity. By enlarging the contract area in the feet, the ultrasonic cleaner can be placed with more stability while the appearance is also improved. Modified mesh style, improve the ultrasonic efficiency of using basket. The amended mesh patterns in baskets enhance the ultrasonic efficacy when baskets are being used.