Mercury(II) sulphate 99,00 AR ACS, Analytical reagent, Rankem®

Supplier: RANKEM
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

M2221 M2222
RANKM2221EA 17968.5 INR
RANKM2221 RANKM2222
Mercury(II) sulphate 99,00 AR ACS, Analytical reagent, Rankem®
Mercury (II) sulphate

High purity reagents that meet or exceed the ACS reagent specifications.


  • Batch-to-batch consistency
  • Suitable for advanced chemical testing
  • Available in various pack size


Description: White crystalline powder

Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850 °C
Density: 6.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now

Specification Test Results

Guarantee Analysis .
Assay (HgSO4) Min. 99.00 %
Identity Passes test
Maximum Limits Of Impurities .
Residue After Reduction Max. 0.02 %
Mercurous Mercury (As Hg) Max. 0.10 %
Chloride (Cl) <=0.003%
Nitrate (NO3) Passes test
Iron (Fe) <=0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR