N,N-Dimethylformamide 99,80 AR ACS, Analytical reagent, Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
D0143
D0144
RANKD0143EA
1029
INR
RANKD0143
RANKD0144
N,N-Dimethylformamide 99,80 AR ACS, Analytical reagent, Rankem®
N,N-Dimethylformamide
High purity analytical reagent that meets or exceeds the ACS reagent specifications.
- Tested for heavy metals
- Suitable for analytical testing
- Available in various pack size
Description: A clear, colourless liquid.
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 °C (1013 hPa) Melting Pt: –60.5 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 57.5 °C (DIN 51755) (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III Merck Index: 13,03269 |
Specification Test Results
Color (APHA) | ≤ 10 |
Guarantee Analysis | . |
Assay (GC) (HCON(CH3)2) | Min. 99.80 % |
Weight Per ml (at 20°C) | 0.948 --> 0.949 g |
Identity (IR) | Conforms |
Maximum Limits Of Impurities | . |
Residue After Evaporation | Max. 0.001 % |
Titrable Acid | Max. 0.0005 meq/g |
Titrable Base | Max. 0.003 meq/g |
Water ( By KF) | Max. 0.10 % |
Copper(Cu) | Max. 0.0005 % |
Iron (Fe) | Max. 0.0005 % |
Lead ( Pb) | Max. 0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...