N,N-Dimethylformamide ≥99.00% LR, Laboratory Reagent, Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
D0141
D0140
D0130
D0146
RANKD0141EA
5401.8
INR
RANKD0141
RANKD0140
RANKD0130
RANKD0146
N,N-Dimethylformamide ≥99.00% LR, Laboratory Reagent, Rankem®
N,N-Dimethylformamide
General lab reagent for routine laboratory use.
- Suitable for routine laboratory usage and classical analysis
- Available in various pack size
Description: A clear, colourless liquid.
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 °C (1013 hPa) Melting Pt: –60.5 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 57.5 °C (DIN 51755) (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3 III Merck Index: 13,03269 |
Specification Test Results
Guarantee Analysis | . |
Assay(Gc) | Min. 99.00 % |
Weight Per Ml (At 20°C) | 0.947 --> 0.949 g |
Refractive Index (At 20°C) | 1.429 --> 1.431 |
Maximum Limits Of Impurities | . |
Acidity | Max. 0.005 % |
Water ( By Kf) | Max. 0.1000 % |
Non-Volatile Matter | Max. 0.00200 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...