Phenol ≥99.50% AR ACS, Analytical reagent (AR), Rankem®
Supplier: RANKEM
|
Danger
|
Synonyms:
Phenylalcohol, Carbolic acid, Hydroxybenzene
P0134
P0136
RANKP0134EA
975.51
INR
RANKP0134
RANKP0136
Phenol ≥99.50% AR ACS, Analytical reagent (AR), Rankem®
Phenol
High purity analytical reagent that meets or exceeds the ACS reagent specifications.
- Batch-to-Batch consistency
- Suitable for analytical testing
Description: Colourless crystalline mass.
Formula:
C₆H₅OH MW: 94.11 g/mol Boiling Pt: 181,8 °C (1013 hPa) Melting Pt: 40,8 °C Density: 1,06 g/cm³ (20 °C) Flash Pt: 81 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002143 CAS Number: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1 II Merck Index: 12,07390 |
Specification Test Results
Guarantee Analysis | . |
Assay (C6H5OH) | Min. 99.50 % |
pH (5% Aqueous Solution) | 4.50 --> 6.00 |
Freezing Point | Min. 40.5 °C |
Identity (IR) | Conforms |
Maximum Limits Of Impurities | . |
Residue After Evaporation | Max. 0.01 % |
Water ( By KF) | Max. 0.50 % |
Tarry Matter | No Reaction |
O.M.P-Cresol | Max. 0.15 % |
Chloride (Cl) | <=0.0005% |
Iron (Fe) | Max. 0.0001 % |
Lead ( Pb) | Max. 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...