Tetrahydrofuran ≥99.5%, Drysolv, Laboratory Reagent, Rankem®
Supplier: RANKEM
|
Danger
|
T2210
T2215
RANKT2210EA
9500
INR
RANKT2210
RANKT2215
Tetrahydrofuran ≥99.5%, Drysolv, Laboratory Reagent, Rankem®
Tetrahydrofuran
General lab reagent for routine laboratory use.
- Suitable for routine laboratory usage and classical analysis
- Controlled moisture and peroxide content
- Stabilised
Description: A clear, colourless liquid with a characteristic odour.
Formula:
C₄H₈O Boiling Pt: 66 °C (1013 hPa) Melting Pt: –108,5 °C Density: 0,888 g/cm³ (20 °C) Flash Pt: −21,5 °C |
MDL Number:
MFCD00005356 CAS Number: 109-99-9 EINECS: 203-726-8 UN: 2056 ADR: 3,II |
Specification Test Results
Guarantee Analysis | . |
Assay(GC) | ≥ 99.50 % |
Weight Per ml (at 20°C) | 0.886 - 0.888 g |
Refractive Index (at 20°C) | 1.407 - 1.408 |
Bht (Stabilizer) | About 250 ppm |
Maximum Limits Of Impurities | . |
Free Acid (As CH3COOH) | ≤ 0.001 % |
Peroxide (As H2O2) | ≤ 0.005 % |
Water ( By KF) | ≤ 0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...