Phenol red sodium salt ACS
Supplier: VWR Chemicals
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide
0648-50GEA
11540
INR
0648-50G
Phenol red sodium salt ACS
Phenol red sodium salt
Formula:
C₁₉H₁₃NaO₅S MW: 376,35 g/mol Melting Pt: 285 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066901 CAS Number: 34487-61-1 EINECS: 252-057-8 Merck Index: 12,07397 |
Specification Test Results
Solubility (0.1%, water) | Pass |
Visual transition interval (pH 6.8 – 8.2) | Pass |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...